missing translation for 'onlineSavingsMsg'
Learn More
Carbazole, 96%, Thermo Scientific Chemicals Product Code.: 10050620

Carbazole, 96%, Thermo Scientific Chemicals

Product Code. 10050620
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10050620

Brand: Thermo Scientific Acros 108260250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-74-8
Molecular Formula C12H9N
Molecular Weight (g/mol) 167.21
MDL Number MFCD00004960
InChI Key UJOBWOGCFQCDNV-UHFFFAOYSA-N
Synonym carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhnShow MoreShow Less
PubChem CID 6854
ChEBI CHEBI:27543
IUPAC Name 9H-carbazole
SMILES N1C2=C(C=CC=C2)C2=C1C=CC=C2

Specifications

Melting Point 240.0°C to 246.0°C
Color Beige-Brown or Beige-Yellow
Boiling Point 355.0°C
Flash Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic bottle
Quantity 25 g
Beilstein 20,433
Merck Index 15,1791
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid- soluble in h2so4,1g/3mL quinoline- 1g/6mL pyridine- 1g/35mL ether,1g/9mL acetone- 1g/120mL benzene,1g/135mL abs. alcohol- slightly soluble in,petroleum ether and
Formula Weight 167.21
Percent Purity 96%
Physical Form Crystalline Powder, Flakes, or Chunks
Chemical Name or Material Carbazole, 96%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0456791 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number108262500,108260050,108260250,108260010
Lot NumberA0448840 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number108262500,108260050,108260250,108260010
Lot NumberA0402148 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number108262500,108260050,108260250,108260010
Lot NumberA0373051 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number108262500,108260050,108260250,108260010
Lot NumberA0445799 Certificate TypeCertificate of Analysis Date28/09/2022 Catalog Number108262500,108260050,108260250,108260010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Carbazole
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbazole, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.