Learn More
Carbazole, 96%, Thermo Scientific Chemicals
23.82 € - 327.63 €
Chemical Identifiers
CAS | 86-74-8 |
---|---|
Molecular Formula | C12H9N |
Molecular Weight (g/mol) | 167.21 |
MDL Number | MFCD00004960 |
InChI Key | UJOBWOGCFQCDNV-UHFFFAOYSA-N |
Synonym | carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn |
PubChem CID | 6854 |
ChEBI | CHEBI:27543 |
IUPAC Name | 9H-carbazole |
SMILES | N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10469780
|
Thermo Scientific Acros
108260050 |
5 g | Glass Bottle |
24.02 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10050620
|
Thermo Scientific Acros
108260250 |
25 g | Plastic bottle |
23.82 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10479780
|
Thermo Scientific Acros
108262500 |
250 g | Plastic bottle |
114.57 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11996591
|
Thermo Scientific Acros
108260010 |
1 kg | Plastic bottle |
327.63 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86-74-8 | |
167.21 | |
UJOBWOGCFQCDNV-UHFFFAOYSA-N | |
6854 | |
9H-carbazole |
C12H9N | |
MFCD00004960 | |
carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn | |
CHEBI:27543 | |
N1C2=C(C=CC=C2)C2=C1C=CC=C2 |
Specifications
86-74-8 | |
Beige-Brown or Beige-Yellow | |
220°C | |
95% min. (GC) | |
MFCD00004960 | |
15,1791 | |
Solubility in water: insoluble. Other solubilities: slightly soluble in acetic acid- soluble in h2so4,1g/3mL quinoline- 1g/6mL pyridine- 1g/35mL ether,1g/9mL acetone- 1g/120mL benzene,1g/135mL abs. alcohol- slightly soluble in,petroleum ether and | |
N1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
167.21 | |
CHEBI:27543 | |
96% | |
Carbazole, 96% |
240.0°C to 246.0°C | |
355.0°C | |
Authentic | |
C12H9N | |
20,433 | |
carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn | |
UJOBWOGCFQCDNV-UHFFFAOYSA-N | |
9H-carbazole | |
6854 | |
167.21 | |
Crystalline Powder, Flakes, or Chunks |
Safety and Handling
GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Suspected of causing cancer.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 201-696-0
missing translation for 'rtecsNumber' : FE3150000
missing translation for 'tsca' : TSCA
- Carbazole
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.