Learn More
Swann-Morton™ Carbon Steel Scalpel Blades
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Brand:  Swann-Morton™ 0101
 View more versions of this product
Product Code. 11738373
  • 84.40 € / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
  • Carbon steel to BS EN 27740
  • Handles and blades sold separately
Specifications
Specifications
Carbon Steel
# 10
The # 10 fits Handles # 3, # 3L, # 3 Graduated, # 5B, # 7, # 9, # B3 and # B3L
BS EN 7740
Scalpel Blade
25 mm
Single-use
10/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products