missing translation for 'onlineSavingsMsg'
Learn More
Carbon tetrabromide, 98%, Thermo Scientific Chemicals Product Code.: 10431121

Carbon tetrabromide, 98%, Thermo Scientific Chemicals

Product Code. 10431121
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10431121

Brand: Thermo Scientific Acros 325151000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-13-4
Molecular Formula CBr4
Molecular Weight (g/mol) 331.64
MDL Number MFCD00000117
InChI Key HJUGFYREWKUQJT-UHFFFAOYSA-N
Synonym carbon tetrabromide, methane, tetrabromo, carbon bromide, methane tetrabromide, bromid uhlicity, carbontetrabromide, methane, tetrabromide, tetrabrommethan, bromid uhlicity czechShow MoreShow Less
PubChem CID 11205
ChEBI CHEBI:47875
IUPAC Name tetrabromomethane
SMILES C(Br)(Br)(Br)Br

Specifications

Melting Point 88°C to 93°C
Color White
Boiling Point 190°C
Assay Percent Range 97.5% min. (On organic basis) (GC)
Packaging Plastic bottle
Linear Formula CBr4
Quantity 100 g
Beilstein 01, 68
Fieser 15,352; 16,366
Solubility Information Solubility in water: insoluble
Formula Weight 331.64
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Carbon tetrabromide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461277 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number325150050,325155000,325151000
Lot NumberA0440219 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number325150050,325155000,325151000
Lot NumberA0451863 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number325150050,325155000,325151000
Lot NumberA0447102 Certificate TypeCertificate of Analysis Date31/10/2022 Catalog Number325150050,325155000,325151000
Lot NumberA0407003 Certificate TypeCertificate of Analysis Date28/03/2022 Catalog Number325150050,325155000,325151000
compliance-icons compliance-icons
Product Identifier
  • Carbon tetrabromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Carbon tetrabromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.