Learn More
Fisherbrand™ Cardboard Cryobox Inserts, 133mm
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
18.95 € - 122.00 €
Specifications
Quantity 10 Pack
Material Cardboard
Product Type Cryo Box Insert
Width (Metric) 133mm
Length (Metric) 133mm
View More Specs
Products 24
Product Code Brand Height (Metric) Array Price Quantity & Availability  
Product Code Brand Height (Metric) Array Price Quantity & Availability  
11842513
View Documents
Fisherbrand™
FB71346
25mm 10 x 10
24.23 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11852513
View Documents
Fisherbrand™
FB71356
30mm 10 x 10
49.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11862513
View Documents
Fisherbrand™
FB71366
40mm 10 x 10
76.54 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11842731
View Documents
Fisherbrand™
59573
40mm 10 x 10
122.00 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11872513
View Documents
Fisherbrand™
FB71376
65mm 10 x 10
29.45 €
Pack of 10
In Stock
to see stock.
Add to basket
11882513
View Documents
Fisherbrand™
FB71349
25mm 16 x 16
44.65 €
Pack of 10
In Stock
to see stock.
Add to basket
11892513
View Documents
Fisherbrand™
FB71359
30mm 16 x 16
44.65 €
Pack of 10
In Stock
to see stock.
Add to basket
11802523
View Documents
Fisherbrand™
FB71369
40mm 16 x 16
50.10 €
Pack of 10
In Stock
to see stock.
Add to basket
11812523
View Documents
Fisherbrand™
FB71379
65mm 16 x 16
44.65 €
Pack of 10
In Stock
to see stock.
Add to basket
11882493
View Documents
Fisherbrand™
FB71340
25mm 4 x 4
88.30 €
Pack of 10
In Stock
to see stock.
Add to basket
11802503
View Documents
Fisherbrand™
FB71360
40mm 4 x 4
36.35 €
Pack of 10
In Stock
to see stock.
Add to basket
11812503
View Documents
Fisherbrand™
FB71370
65mm 4 x 4
37.95 €
Pack of 10
In Stock
to see stock.
Add to basket
11557693
View Documents
Fisherbrand™
FB71343
25mm 7 x 7
23.30 €
Pack of 10
In Stock
to see stock.
Add to basket
11517703
View Documents
Fisherbrand™
FB71353
30mm 7 x 7
18.95 €
Pack of 10
In Stock
to see stock.
Add to basket
11577703
View Documents
Fisherbrand™
FB71363
40mm 7 x 7
28.75 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11527703
View Documents
Fisherbrand™
FB71354
30mm 8 x 8
19.55 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11882503
View Documents
Fisherbrand™
FB71364
40mm 8 x 8
49.70 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11892503
View Documents
Fisherbrand™
FB71374
65mm 8 x 8
29.45 €
Pack of 10
In Stock
to see stock.
Add to basket
11802513
View Documents
Fisherbrand™
FB71345
25mm 9 x 9
64.12 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11812513
View Documents
Fisherbrand™
FB71355
30mm 9 x 9
50.60 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
11832513
View Documents
Fisherbrand™
FB71375
65mm 9 x 9
30.40 €
Pack of 10
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
11822513
View Documents
Fisherbrand™
FB71365
40mm 9 x 9
49.10 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10084213
View Documents
Fisherbrand™
FB71344
25mm 8 x 8
23.30 €
Pack of 10
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
12681565
View Documents
Fisherbrand™
FB71373
65mm 7 x 7
29.90 €
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Ultra low temperature-resistant cardboard
  • For sample storage
  • Temperature-resistant down to -80°C
  • Water-repellent finish
Specifications
Specifications
10 Pack
Cryo Box Insert
133mm
Cardboard
133mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Cardboard Cryobox Inserts, 133mm