Learn More
Fisherbrand™ Cardboard Cryobox Inserts, 136mm
Combine these inserts with Cryoboxes for flexible cell storage and sample handling. Fisherbrand™ Cardboard Cryobox Inserts feature ultra-low temperature resistant cardboard with water repellent finish for sample storage.
16.80 € - 89.45 €
Product Code | Brand | Height (Metric) | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Array | Price | Quantity & Availability | ||||
11507683
|
Fisherbrand™
|
30mm | 10 x 10 |
16.80 €
Pack of 10 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10354003
|
Fisherbrand™
|
40mm | 10 x 10 |
57.40 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10797093
|
Fisherbrand™
|
65mm | 10 x 10 |
25.10 €
Pack of 10 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||||
10008872
|
Fisherbrand™
|
25mm | 9 x 9 |
21.05 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10384573
|
Fisherbrand™
|
40mm | 9 x 9 |
26.85 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10130033
|
Fisherbrand™
|
65mm | 9 x 9 |
72.15 €
Pack of 10 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||||
10767093
|
Fisherbrand™
|
65mm | 8 x 8 |
25.90 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10232963
|
Fisherbrand™
|
65mm | 7 x 7 |
71.25 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10130903
|
Fisherbrand™
|
25mm | 4 x 4 |
35.45 €
Pack of 10 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
10314633
|
Fisherbrand™
|
30mm | 4 x 4 |
35.45 €
Pack of 10 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||||
10394053
|
Fisherbrand™
|
40mm | 4 x 4 |
35.45 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
10465703
|
Fisherbrand™
|
65mm | 4 x 4 |
89.45 €
Pack of 10 |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||||
11872523
|
Fisherbrand™
|
40mm | 7 x 7 |
66.40 €
Pack of 10 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||||
11892523
|
Fisherbrand™
|
25mm | 8 x 8 |
21.05 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11547673
|
Fisherbrand™
|
25mm | 10 x 10 |
24.41 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11826893
|
Fisherbrand™
|
25mm | 16 x 16 |
45.20 €
Pack of 10 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11862523
|
Fisherbrand™
|
30mm | 7 x 7 |
17.90 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11802533
|
Fisherbrand™
|
20mm | 8 x 8 |
19.05 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11812533
|
Fisherbrand™
|
40mm | 8 x 8 |
56.00 €
Pack of 10 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11577683
|
Fisherbrand™
|
40mm | 16 x 16 |
43.55 €
Pack of 10 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
||||
11816913
|
Fisherbrand™
|
65mm | 16 x 16 |
43.55 €
Pack of 10 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
11932421
|
Fisherbrand™
|
30mm | 9 x 9 |
19.05 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11836893
|
Fisherbrand™
|
50mm | 16 x 16 |
43.55 €
Pack of 10 |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
Description
- Ultra low temperature-resistant cardboard
- For sample storage
- Temperature-resistant down to -80°C
- Water-repellent finish