Learn More
Carl Zeiss™ Immersol™ Immersion Oil
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
Brand:  Carl Zeiss™ 444960-0000-000
 View more versions of this product
Product Code. 10539438
  • 103.00 € / 20mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.
  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss
Specifications
Specifications
Immersol 518 F
Oiler
1.518
ISO 8036-1/2
20 mL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Immersol 518 F
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
Supplemental information
  • MIXTURE LIST-Contains : Adipic acid-di(8-methyl-tricyclo(5.2. 1.0.2.6.) decane) ester , Benzyl benzoate