missing translation for 'onlineSavingsMsg'
Learn More

Carl Zeiss™ Immersol™ Immersion Oil

Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.

46.65 € - 103.00 €

Specifications

Refractive Index 1.518
View More Specs

Products 2
Product Code Brand Certifications/Compliance Quantity Packaging Type Type Price Quantity & Availability  
Product Code Brand Certifications/Compliance Quantity Packaging Type Type Price Quantity & Availability  
10539438
View Documents
Carl Zeiss™
444960-0000-000
ISO 8036-1/2 20 mL Oiler Immersol 518 F
103.00 €
20mL
In Stock
to see stock.
Add to basket
 
10418653
View Documents
Carl Zeiss™
000000-1111-806
ISO 8036-1 20 mL Oiler Immersol 518 N
46.65 €
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
Description

Description

Increases the resolving power of a microscope. Zeiss Immersol™ is a transparent oil with a refractive index of 1.518. It is free of fluorescence and halogen, and is certified according to ISO 8036-1/2.

  • Bubble-free immersion oil
  • Suitable for oil immersion objectives from Carl Zeiss

Specifications

Specifications

1.518
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Immersol 518 N

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Adipic acid-di(8-methyl-tricyclo(5.2. 1.0.2.6.) decane) ester , Bis (isopropyl)naphthalene

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.