Learn More
Motic™ CCIS™ Plan Achromatic Phase Objective
Provides a flat field of view while also correcting for color aberration. Motic™ CCIS™ Plan Achromatic Phase Objective is useful on specimens that do not absorb light.
Brand:  Motic™ 1101001703091
 View more versions of this product
Product Code. 11762348
  • 193.00 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with AE31 Inverted Microscope
  • Achromatic Plan Phase Objective
  • Flat field
  • Eliminates color aberration
  • Magnification and Numerical Apertures available:
    • 10X/0.25 (WD=4.1mm) Ph1 for AE
    • 4X/0.10 (WD=12.6mm) Ph0 for AE
Specifications
Specifications
PL Ph1 10X
0.25 mm, WD=4.1 mm
AE31 inverted microscope
1 Piece
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ CCIS™ Plan Achromatic Phase Objective > Magnification Power: 10X; Focus Distance: 0.25mm