missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh
Used as cheese, food additive and binder for safety matches.
Brand: MP Biomedicals™ 0290452025
This item is not returnable.
View return policy
Description
- Phosphoprotein
- Extracted from bovine milk
- Used as food source
- CAS: 9000-71-9

Specifications
Casein | |
Powder | |
25 lb. |
Yellow | |
Used as cheese, food additive and binder for safety matches | |
Store at room temperature (15°C to 30°C) |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh > Quantity: 25lb
Spot an opportunity for improvement?Share a Content Correction