missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Catechol Violet

46.83 € - 173.87 €

Chemical Identifiers

CAS 115-41-3
Molecular Formula C19H14O7S
Molecular Weight (g/mol) 386.37
MDL Number MFCD00005868
InChI Key RRRCKIRSVQAAAS-UHFFFAOYSA-N
Synonym Pyrocatechol sulfonephthalein; Pyrocatechol Violet
PubChem CID 66993
IUPAC Name 4-[3-(3,4-dihydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]benzene-1,2-diol
SMILES OC1=CC=C(C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C(O)=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11455517
SDS View Documents
Thermo Scientific Chemicals
B23513.06
5 g
46.83 €
5g
Please to purchase this item. Need a web account? Register with us today!
11465517
SDS View Documents
Thermo Scientific Chemicals
B23513.14
25 g
173.87 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catechol Violet is used as a complexometric indicator dye. It inhibits the synthesis of globin in reticulocyte lysates. It is also used as a pH indicator with a transition interval from 0 to 8.

Solubility
Soluble in water, ethylene glycol, monomethyl ether and ethanol.

Notes
Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P261-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c

H314-H335

missing translation for 'einecsNumber' : 204-088-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Catechol Violet

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.