Learn More
CCS Viscosity Standard, ASTM D 5293 and SAE Specification J314, Paragon Scientific™
Paragon ISO 17025 / ISO 17034 certified CCS viscosity standards are for the calibration and verification of analytical equipment used in Cold Cranking Simulator (CCS) testing to ASTM D5293 and SAE Specification J300.
Brand: Paragon Scientific CL22
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Manufactured in the United Kingdom
- 2 year shelf life
Specifications
Glass Amber Bottle | |
500 mL | |
Yes | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Nominal values quoted. Certified Values will vary from batch to batch.