missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD20 Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications.

Brand:  Epredia™ MS-340-R7

 View more versions of this product

Product Code. 12621067

  • 270.00 € / 7mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Mouse

Clone: L26

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with cow, dog, pig and rat.

Epitope: Cytoplasmic domain

Immunogen: Human tonsil B cells

Molecular Weight: 30-33kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
Specifications

Specifications

CD20 Ab-1
Monoclonal
Unconjugated
Human tonsil B cells
IVD
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin)
L26
Mouse
7 mL
B-Cell
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.