missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD20 Ab-1, Mouse Monoclonal Antibody

Obtain accurate and reproducible results in immunohistochemistry, immunofluorescence and flow cytometry applications.

142.00 € - 610.00 €

Specifications

Antigen CD20 Ab-1
Clone L26
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Form Quantity Price Quantity & Availability  
Product Code Brand Form Quantity Price Quantity & Availability  
12651067
View Documents
Epredia™
MS-340-S0
Concentrated 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12671067
View Documents
Epredia™
MS-340-S1
Concentrated 500 μL
399.00 €
500µL
Please to purchase this item. Need a web account? Register with us today!
12641067
View Documents
Epredia™
MS-340-S
Concentrated 1 mL
610.00 €
1mL
Please to purchase this item. Need a web account? Register with us today!
12621067
View Documents
Epredia™
MS-340-R7
Prediluted 7 mL
270.00 €
7mL
Please to purchase this item. Need a web account? Register with us today!
12631067
View Documents
Epredia™
MS-340-RQ
Ready-to-Use 12 mL
434.00 €
12mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD20 is a non-Ig differentiation antigen of B cells and the expression of CD20 is restricted to normal and neoplastic B cells, being absent from all other leukocytes and tissues. It acts as calcium channel involved in B cell activation and cell cycle progression.

Host Species: Mouse

Clone: L26

Isotype: IgG2a/κ

Species Reactivity: Human. Does not react with cow, dog, pig and rat.

Epitope: Cytoplasmic domain

Immunogen: Human tonsil B cells

Molecular Weight: 30-33kDa

Positive Control: Tonsil

Cellular Localization: Membrane/cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry

Specifications

Specifications

CD20 Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human tonsil B cells
L26
Monoclonal
Mouse
B-Cell
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.