Learn More
CD4 Mouse anti-Human , PE, Clone: SK3 (also known as Leu3a), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566910
224.70 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
CD4 Monoclonal antibody specifically detects CD4 in Human samples. It is validated for Flow Cytometry
Specifications
CD4 | |
Monoclonal | |
PE | |
L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55) | |
Human Peripheral Blood T Cells | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
SK3 (also known as Leu3a) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.