missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD44v6 Mouse, Clone: CD44v6/1246, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 960-MSM7-P0
This item is not returnable.
View return policy
Specifications
CD44v6 | |
Cell surface | |
CD44v6/1246 | |
Unconjugated | |
XP_016874073 | |
Mouse | |
80-95kDa | |
Protein A/G Purified | |
RUO | |
960 | |
IgG2a κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CD44 | |
Recombinant fragment corresponding to the v6 domain of human CD44 | |
Cervix or Tongue Squamous Cell Carcinoma | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |