missing translation for 'onlineSavingsMsg'
Learn More

CD44v6 Mouse, Clone: CD44v6/1246, enQuireBio™

Mouse Monoclonal Antibody

199.00 € - 485.00 €

Specifications

Antigen CD44v6
Clone CD44v6/1246
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15951258
View Documents Promotion Details
EnquireBio
960-MSM7-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15961258
View Documents Promotion Details
EnquireBio
960-MSM7-P1
100 μg
485.00 €
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD44v6
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
960
Recombinant fragment corresponding to the v6 domain of human CD44
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
80-95kDa
CD44v6/1246
Immunohistochemistry
Unconjugated
RUO
XP_016874073
CD44
IgG2a κ
Protein A/G Purified
Cell surface
Cervix or Tongue Squamous Cell Carcinoma
Videos
SDS
Documents

Documents

Product Certifications