Learn More
Vileda Professional™ CE Sweeper and Pan™
The CE Sweeper and Pan™ is ideal for ergonomic and efficient liquid removal. The system is hygienic, lightweight, and flexible. Perfect for dry or damp conditions.
Brand:  Vileda Professional™ 119914
263.61 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17862476
  • 316.00 € / Pack of 5
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sweep up broken glass in your cleanroom quickly and easily
  • No need to change gloves when an item is dropped on the floor
  • Keeps workers from touching the floor within the controlled environment
  • Can be sprayed or wiped down with common disinfectants
  • Stands up easily for storage or can be hung
  • Adjustable bag holder for different size collection bags
Specifications
Specifications
Open Lobby Dustpan
For ergonomic and efficient dirt and dust removal directly into the bag
Janitorial
Metal/Plastic