missing translation for 'onlineSavingsMsg'
Learn More
Celastrol, Tocris Bioscience™ Product Code.: 18712754

Celastrol, Tocris Bioscience™

Product Code. 18712754
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18712754

Brand: Tocris Bioscience 3203/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibits TNF-α-induced NF-κB activation

Antioxidant and anti-inflammatory agent. Potently inhibits lipid peroxidation in mitochondria and inhibits TNF-α-induced NFκB activation. Also shown to inhibit topoisomerase II activity in vitro (IC50 = 7.41 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34157-83-0
Molecular Formula C29H38O4
Molecular Weight (g/mol) 450.62
MDL Number MFCD03424073
InChI Key KQJSQWZMSAGSHN-JJWQIEBTSA-N
Synonym celastrol, tripterine, tripterin, 2r,4as,6as,12br,14as,14br-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acid, triterpene, celastrol, celastrus scandens, unii-l8gg98663l, 3-hydroxy-24-nor-2-oxo-1 10 ,3,5,7-friedelatetraen-29-oic acid, 2r,4as,6ar,6as,14as,14br-10-hydroxy-2,4a,6a,6a,9,14a-hexamethyl-11-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid, d:a-friedo-24-noroleana-1 10 ,3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo-, 20alphaShow MoreShow Less
PubChem CID 122724
ChEBI CHEBI:63959
IUPAC Name (2R,4aS,6aS,12bR,14aS,14bR)-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydropicene-2-carboxylic acidShow MoreShow Less
SMILES CC1=C(O)C(=O)C=C2C1=CC=C1[C@@]2(C)CC[C@@]2(C)[C@@H]3C[C@@](C)(CC[C@]3(C)CC[C@]12C)C(O)=O

Specifications

Quantity 50 mg
Formula Weight 450.61
Percent Purity >99%
Chemical Name or Material Celastrol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Celastrol, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.