missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Cell Cryopreservation Medium with 10% DMSO
Balanced, protein free, ready to use cryopreservation solution with 10% USP DMSO (CryoPres™).
Brand: MP Biomedicals™ 092780248
This item is not returnable.
View return policy
Description
- Cell Cryopreservation Medium with 10% DMSO is a balanced, protein free, ready-to-use cryopreservation solution with 10% USP DMSO (Cryopres™)
- It does not require any dilution or further processing and can be used for cells cultured in serum free medium or with serum containing medium

Specifications
Stored at 2 to 8°C | |
Sterile | |
50 mL |
Liquid | |
Cryopreservation |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Cell Cryopreservation Medium with 10% DMSO
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dimethyl Sulfoxide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ Cell Cryopreservation Medium with 10% DMSO > Quantity: 50 mL
Spot an opportunity for improvement?Share a Content Correction