Learn More
Gibco™ DPBS, no calcium, no magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Marque: Gibco™ 14190250
186.89 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
This DPBS is modified as follows:
Without: Calcium, Magnesium, Phenol Red
Informations sur la commande
Shipping Condition: Room Temperature

Spécification
No Sodium Pyruvate | |
Clear | |
Bottle | |
Gibco™ | |
10 x 500 mL | |
Mammalian Cell Culture | |
7.0 to 7.3 | |
Room Temperature | |
Liquid |
Animal Origin-Free | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
|
1 X | |
DPBS | |
Dulbecco's Phosphate Buffered Saline | |
270 - 300 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Suggestions de produits
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2534474 | Certificate of Analysis | 21/02/2025 | 14190359,14190342,14190144,14190367,14190250 |
2534480 | Certificate of Analysis | 21/02/2025 | 14190359,14190144,14190342,14190367,14190250 |
2482073 | Certificate of Analysis | 21/02/2025 | 14190359,14190144,14190342,14190367,14190250 |
2530913 | Certificate of Analysis | 21/02/2025 | 14190359,14190342,14190144,14190367,14190250 |
2446697 | Certificate of Analysis | 21/02/2025 | 14190359,14190144,14190342,14190367,14190250 |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.