missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ cellgro™ DMEM/Ham's F-12 Mix
Products undergo a variety of tests to determine functionality and toxicity using growth-promotion analysis. Corning™ cellgro™ DMEM/Ham's F-12 Mix support cell culture growth and substitution of Pyridoxine HCl for Pyridoxal ensures improved stability over original formulation.
153.19 € - 301.64 €
Specifications
Content And Storage | 2 to 8°C |
---|---|
Concentration | 1 X |
Form | Liquid |
Product Type | DMEM and Ham's F-12, 50/50 Mix |
Endotoxin Level | <0.25EU/mL |
Product Code | Brand | Quantity | Shelf Life | With Additives | Without Additives | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Shelf Life | With Additives | Without Additives | Price | Quantity & Availability | ||
15323611
|
Corning™
10-092-CM |
6 x 1 L | 12 Months | Glutamine, HEPES | None |
247.95 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15323561
|
Corning™
15-090-CV |
6 x 500 mL | 18 Months | Glutamine | None |
153.19 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15313611
|
Corning™
10-090-CM |
6 x 1 L | 12 Months | Glutamine | None |
301.64 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15353561
|
Corning™
16-405-CV |
6 x 500 mL | 12 Months | Glutamine | No Phenol Red |
196.55 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Modification of Basal Medium Eagle (BME) containing four-fold concentrations of the amino acids and vitamins found in BME
- Original formulation was used to culture embryonic mouse cells
- Has been modified to support variety of normal and transformed cells, such as primary cultures of mouse and chicken cells
- Formulations can differ in amount or absence of components such as glucose, sodium pyruvate, sodium bicarbonate and L-glutamine
- Tested to determine pH and osmolality
- Low levels of endotoxin, and absence of contamination
- Tested for bacterial endotoxin using the Limulus Amoebocyte Lysate (LAL) Chromogenic Assay
- Ham's F-12 media is based on Ham's F-10 medium with increased concentrations of choline, inositol, putrescine, and several amino acids
- Mycoplasma: Tested Negative
- Sterility: Pass
Specifications
2 to 8°C | |
Liquid | |
<0.25EU/mL |
1 X | |
DMEM and Ham's F-12, 50/50 Mix | |
To assess growth promotion and cytotoxicity characteristics |
Safety and Handling
missing translation for 'shelfLife' : 12 Months