missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform
Rapidly perform high content imaging assays with this affordable, compact HCS platform.
Brand: Thermo Scientific™ CX51110
This item is not returnable.
View return policy
Description
Rapidly perform high content imaging assays with the affordable, compact Thermo Scientific™ CellInsight™ CX5 High Content Screening (HCS) Platform.
The CellInsight CX5 High Content Screening (HCS) Platform is a fast, simple-to operate, automated cellular imaging and analysis benchtop platform designed for quantitative imaging. For the researcher looking to gain the advantage of quantitative cell analysis for the first time or for those who are looking to increase scale and capacity, the CellInsight CX5 HCS Platform offers a costs effective, but powerful way to quickly automate quantitative cellular imaging.Highlights:
- Flexible – designed to work with or without fluorescent labeling, the new CellInsight CX5 HCS Platform uses transmitted light allowing users to explore more cell biology without the restriction of fluorescent dyes or to use the full capabilities of a five-color light engine
- Affordable – automated high content imaging for all budgets
- Easy to Use – quickly build and optimize image-based assays with included Thermo Scientific™HCS Studio™ 2.0 Cell Analysis Software
- Performance – scan and analyze a 96-well plate in under four minutes
- Quality – high-quality imaging and virtually maintenance-free operation
- Compact – fits on a bench and is less than 2 ft. on all sides
Capabilities:
- Make individual cell, subcellular and multicellular measurements
- Save time by automatically collecting only the images and data you need to be statistically relevant
- Make multiple measurements per object, over time if desired
- Measure subpopulations and reject undesired objects automatically
Easy to Use Applications:
- Comprehensively pre-validated and user configurable image analysis without scripting or programming
- Exploratory assay development tools and specific biology optimized tools
- Define biological thresholds for outputs based on control populations
- Ability to create multi-parameter “gates” from individual output measurements
Hardware and Software:
- HCS Studio 2.0 Cell Analysis Software – the premier high content instrument control, visualization and analysis program for novice to advanced users featuring image/data portability
- Ultra-stable, long-life light source reduces variability, increases data quality, and decreases operating costs
- Fluorescence options which include near-IR wavelengths
- Proprietary Thermo Scientific™ AccuFocus System is fast, yet flexible, imaging cells with high-fidelity every time
- Unique intelligent acQuisition (iQ) real time analysis feedback to optimize image collection and data quality
- Compatible with SBS standard microplates (6-1536 well) and slides
Compatible with: Optional Automation – Thermo Scientific™ Orbitor™ RS Plate Mover and other Thermo Scientific™ Laboratory Automation Solutions; Optional Modules - Thermo Scientific™ High Content Modules
Specifications
CellInsight | |
Thermo Scientific™ Orbitor™ RS Plate Mover | |
Brightfield, Fluorescence | |
High-throughput Compatible, Multiplexing, Automated Protocols | |
Additional Objectives Available, Low and High NA, Single Position 2x-20x | |
JPG Files, TIFF Files, PNG Files, C01, BMP, BIN, PZFX, ICE, GIF Files, DIB, CSV, FCS 3.1, XLSX | |
8GB RAM | |
Windows 7™ 64-bit PC with quad core 3.7GHz processor, 8GB RAM, and 500GB Hard Drive | |
Permissible ambient temperature range during operation : + 18 deg C to + 25 deg C, Permissible relative humidity during operation (noncondensing): > 75% at 30 deg C | |
45 kg | |
120V |
CeLLInsight™ CX5 High Content Screening Platform with HCS Studio Cell Analysis Software and Store Express | |
Green, Blue, Near-Infrared, Far-Red, Red | |
Slide(s), 96-well plate, 384-well plate, 6-well plate, 1536-well plate, 8 wells, 24-well plate | |
740⁄809, 560⁄607, 485⁄521, 386⁄440, 650⁄694 | |
CCD Camera, LED, solid-state 5-color light engine | |
24in High-resolution widescreen monitor | |
Windows 7 | |
Thermo Scientific™ Store™ Image and Database Management Software, Thermo Scientific™ HCS Studio™ Cell Analysis Software | |
41.4 cm (16.3 in) (W) x 37.2 cm (14.6 in) (H) x 59.4 cm (23.4 in) (D) | |
1 each | |
300 W |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction