missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cells-to-CT™ Bulk Fast Advanced RT Reagents
GREENER_CHOICE

Invitrogen Fast Advanced Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription (RT) in the TaqMan Fast Advanced Cells-to-CT Kit and the SYBR Green Fast Advanced Cells-to-CT Kit.

Brand:  Invitrogen™ A39110

Product Code. 16396542

  • 9870.00 € / 2500 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Cells-to-Ct 20X Fast Advanced RT enzyme mix and 2X RT buffer
This item is not returnable. View return policy

Description

Description

Invitrogen Fast Advanced Cells-to-CT Bulk RT Reagents are separately-available, large-quantity versions of the reagents used for reverse transcription (RT) in the TaqMan Fast Advanced Cells-to-CT Kit and the SYBR Green Fast Advanced Cells-to-CT Kit (20X RT enzyme mix and 20X RT buffer).

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 6.8 mL 20X Fast Advanced RT Enzyme Mix
• 68 mL 2X Fast Advanced RT Buffer

Store at -5° to -30°C.
High
Fewer resources used and less waste, Less hazardous
Cells-to-CT™
2500 reactions
2500 Reactions
20X
2500 Reactions
Cell
Primer-probe
Fast
High-throughput Compatible
Bulk Fast Advanced Rt Reagent
Dry Ice
Gene Expression
20X
Lysate
68 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2801875 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA39110
Lot Number2822532 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA39110
Lot Number2792212 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA39110
Lot Number2830494 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA39110
Lot Number2808549 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA39110
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Cells-to-CT™ Bulk Fast Advanced RT Reagents > 2500 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.