missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry

CellTrace™ CFSE Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

143.00 € - 311.00 €

Specifications

Detection Method Fluorescence
For Use With (Equipment) Flow Cytometer
Product Type Cell Proliferation Kit
Emission 488 nm
Format Tube(s)
View More Specs

Products 2
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
15598431
View Documents
Invitrogen™
C34570
20 Reactions
143.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
10013162
View Documents
Invitrogen™
C34554
180 Reactions
311.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Fluorescence
Cell Proliferation Kit
Tube(s)
Room Temperature
Flow Cytometer
488 nm
CellTrace™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.