missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry

CellTrace™ CFSE Cell Proliferation Kit is used for in vitro and in vivo labeling of cells to trace multiple generations using dye dilution by flow cytometry.

Brand:  Invitrogen™ C34570

133.71 EUR valid until 2025-12-31
Use promo code "24276" to get your promotional price.


 View more versions of this product

Product Code. 15598431

  • 143.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Contains 1 vial of CellTrace CFSE (lyophilized powder) and 1 vial of DMSO.

Store in freezer (-5°C to -30°C) and protect from light.
Fluorescence
Flow Cytometer
CellTrace™ CFSE
Solid
CellTrace™
CFSE & Related Compounds
20 Reactions
CellTrace™ Cell Proliferation Kit, 20 rxns, CFSE, Solid, Fluorescent Detection Method, 492/517nm Excitation/Emission Wavelength, CellTrace™ CFSE Label/Dye, –5 to –30°C Storage, Use with Flow Cytometer
Proliferation Assay
Cell Proliferation Kit
488 nm
Tube(s)
20 reactions kit
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ CellTrace™ CFSE Cell Proliferation Kit, for flow cytometry > 20 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.