Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Minimizes the hands-on time involved in extraction. Macherey Nagel™ Cellulose Extraction Thimbles are used for holding solid materials from which substances are to be extracted with a suitable solvent.
Specifications
Quantity 25 Pk.
Material Cellulose
For Use With (Application) Analysis of soil contaminants, Analysis of soil contaminants
Product Type Extraction Thimbles
View More Specs
Products 4
Product Code Brand Diameter (Metric) Inside Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inside Height (Metric) Price Quantity & Availability  
10620303
View Documents Promotion Details
Macherey-Nagel™
645006
22 mm 80 mm N/A
N/A
10022751
View Documents Promotion Details
Macherey-Nagel™
645024
31 mm 118 mm N/A
N/A
10422032
View Documents Promotion Details
Macherey-Nagel™
645025
31 mm 180 mm N/A
N/A
10536101
View Documents Promotion Details
Macherey-Nagel™
645004
15 mm 100 mm N/A
N/A
Description
Description
  • Manufactured from pure cellulose
  • Used in the fields of air and waste gas analysis for collecting solid particles
  • The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
25 Pk.
Cellulose
Analysis of soil contaminants, Analysis of soil contaminants
Extraction Thimbles