missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Cellulose Sani-Stick with Letheen Broth
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
Brand: LABPLAS KSS-67315-LT
This item is not returnable.
View return policy
Description
- Cellulose Sani-Stick
- Saturated with Letheen broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
Gloves |