missing translation for 'onlineSavingsMsg'
Learn More

LABPLAS Cellulose Sani-Stick with Letheen Broth

Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.

271.00 € - 326.00 €

Specifications

Product Type Hygiene Sponge Sampling Kit
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17986499
View Documents
LABPLAS
KSS-67310-LT
None
271.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
17936689
View Documents
LABPLAS
KSS-67315-LT
Gloves
326.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Cellulose Sani-Stick
  • Saturated with Letheen broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications

Specifications

Hygiene Sponge Sampling Kit
Videos
SDS
Documents

Documents

Product Certifications