missing translation for 'onlineSavingsMsg'
Learn More

Scotch-Brite™ Cellulose Sponge 799

Dual function sponge with a white pad for light scouring on any delicate surface in kitchens, bathrooms, restaurants, etc.

Brand:  Scotch-Brite™ 7100157793

Product Code. 17198770

  • 955.00 € / Pack of 240
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Cellulose sponge with fiber fleece for cleaning sensitive surfaces
  • Long lasting due to abrasive particles spread throughout the pad
  • Can withstand common cleaning agents
  • Dual functioning sponge and scouring web
  • Non-rusting and resilient
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sponges
131 x 88 mm
131 mm
Dual Functioning
Pink, White
Cleaning Sensitive Surfaces
Cellulose
88 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Scotch-Brite™ Cellulose Sponge 799 > 131 x 88 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.