missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose triacetate

55.17 € - 97.08 €

Chemical Identifiers

CAS 9012-09-3
Molecular Formula (C24H32O16)n
MDL Number MFCD00132680
InChI Key NNLVGZFZQQXQNW-ADJNRHBOSA-N
Synonym triacetylcellulose, cellulose triacetate, beta-cellotriose undecaacetate, 2r,3r,4s,5r,6s-4,5-bis acetyloxy-6-2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-acetyloxy methyl oxan-3-yl oxy-3-2s,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate
PubChem CID 44263853
IUPAC Name [(2R,3R,4S,5R,6S)-4,5-diacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-6-[(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-2-yl]methyl acetate
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10489650
View Documents
Thermo Scientific Chemicals
177822500
250 g Plastic bottle
55.17 €
250g
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
10124340
View Documents
Thermo Scientific Chemicals
177825000
500 g Plastic bottle
97.08 €
500g
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9012-09-3
MFCD00132680
triacetylcellulose, cellulose triacetate, beta-cellotriose undecaacetate, 2r,3r,4s,5r,6s-4,5-bis acetyloxy-6-2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-acetyloxy methyl oxan-3-yl oxy-3-2s,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate
[(2R,3R,4S,5R,6S)-4,5-diacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-6-[(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-2-yl]methyl acetate
(C24H32O16)n
NNLVGZFZQQXQNW-ADJNRHBOSA-N
44263853

Specifications

White to Yellow
Pellets or Rods
MFCD00132680
triacetylcellulose, cellulose triacetate, beta-cellotriose undecaacetate, 2r,3r,4s,5r,6s-4,5-bis acetyloxy-6-2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-acetyloxy methyl oxan-3-yl oxy-3-2s,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate
NNLVGZFZQQXQNW-ADJNRHBOSA-N
[(2R,3R,4S,5R,6S)-4,5-diacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-6-[(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-2-yl]methyl acetate
Authentic
9012-09-3
(C24H32O16)n
10, 1926
Solubility in water: .. Other solubilities: soluble in chloroform, methylene dichloride and, tetrachloroethane
CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC3C(C(C(C(O3)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
44263853
Cellulose triacetate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : TSCA

Product Identifier
  • Cellulose triacetate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only