Learn More
OHAUS™ Cent-O-Gram™ and Dial-O-Gram™ Balances
Support centigram accuracy and encourage student precision. OHAUS Cent-O-Gram and Dial-O-Gram Balance comes with three-point base, special floating bearing principle, and beam design which eliminates need for level adjustment.
Brand:  OHAUS™ 80000010
249.30 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 10478861
  • 344.00 € / Each
Explore more special offers
Description
Description
  • Quick zeroing by means of a zero-adjust knob at beam end
  • Built-in specific gravity platform recessed in the base
  • Stainless-steel weighing pans and bearing covers prevent rust and corrosion
  • Precision-ground knife edges and self-aligning agate bearings provide maximum sensitivity and reproducibility
  • Notched beams provide positive positioning of poise
  • Magnetic damping slows oscillation
Order Info
The Dial-O-Gram (S40027) model replaces the two front beams with a vernier dial for faster reading results.
Services
Five years
TRUSTED_SUSTAINABILITY
Specifications
Specifications
3.5 in.
7 in.
0.27 in.
0.7 cm
22.86 cm
Stainless Steel
35.56 cm
0.6 lb., 300 g
9 cm
1 x 0.01g (front beam), 10 x 1 (second beam), 100
17.78 cm
9 in.
311-00
14 in.
5 Years
Cent-O-Gram™ Balance
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Cent-O-Gram™ and Dial-O-Gram™ Balances > Without Vernier dial