Learn More
Lenz Laborglasinstrumente™ Distilling Links
Distilling Links for the connection of distillation thermometers of 55 mm built-in length.
Brand: Lenz Laborglasinstrumente™ 05053414
Description
- With Liebig condenser
- Outlet with NS cone
- Water connection with glass olives, with NS socket
Specifications
For distillation thermometers of 55 mm built-In length | |
Duran Glass | |
400 mm |
Distilling Link | |
NS 29/32 |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.