Learn More
Cerium(III) acetate sesquihydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals
53.57 € - 726.62 €
Chemical Identifiers
CAS | 537-00-8 |
---|---|
Molecular Formula | C6H9CeO6 |
Molecular Weight (g/mol) | 317.248 |
MDL Number | MFCD00051060 |
InChI Key | VGBWDOLBWVJTRZ-UHFFFAOYSA-K |
Synonym | cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate |
PubChem CID | 10826 |
IUPAC Name | cerium(3+);triacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361079
|
Thermo Scientific Alfa Aesar
011324.09 |
10 g |
53.57 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371079
|
Thermo Scientific Alfa Aesar
011324.18 |
50 g |
229.14 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11351079
|
Thermo Scientific Alfa Aesar
011324.30 |
250 g |
726.62 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Specialty chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
537-00-8 | |
317.248 | |
VGBWDOLBWVJTRZ-UHFFFAOYSA-K | |
10826 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3] |
C6H9CeO6 | |
MFCD00051060 | |
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate | |
cerium(3+);triacetate |
Specifications
308°C (decomposition) | |
Crystalline Aggregates | |
MFCD00051060 | |
Soluble in water (265g/L) at 15°C. | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Ce+3] | |
317.248 | |
344.28 (317.26 Anhydrous) | |
Reacton™ | |
Cerium(III) acetate sesquihydrate |
537-00-8 | |
C6H9CeO6 | |
cerium acetate, cerous acetate, cerium 3+ acetate, cerium iii acetate, cerium triacetate, cerium 3+ triacetate, unii-yyb5291r2h, acetic acid, cerium 3+ salt, cerium iii acetate sesquihydrate | |
VGBWDOLBWVJTRZ-UHFFFAOYSA-K | |
cerium(3+);triacetate | |
10826 | |
99.99% | |
(REO) |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
missing translation for 'einecsNumber' : 208-654-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only