Learn More
Cerium(III) oxalate hydrate, 99%, Thermo Scientific Chemicals
84.42 € - 261.30 €
Chemical Identifiers
CAS | 15750-47-7 |
---|---|
Molecular Formula | C6Ce2O12 |
Molecular Weight (g/mol) | 544.29 |
MDL Number | MFCD00013082 |
InChI Key | ZMZNLKYXLARXFY-UHFFFAOYSA-H |
Synonym | cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate |
PubChem CID | 91998046 |
IUPAC Name | cerium;oxalic acid;hydrate |
SMILES | [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329387
|
Thermo Scientific Alfa Aesar
040227.14 |
25 g |
84.42 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319387
|
Thermo Scientific Alfa Aesar
040227.22 |
100 g |
261.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in ceramics, glass, phosphors and pyrotechnics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15750-47-7 | |
544.29 | |
ZMZNLKYXLARXFY-UHFFFAOYSA-H | |
91998046 | |
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O |
C6Ce2O12 | |
MFCD00013082 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
cerium;oxalic acid;hydrate |
Specifications
(decomposition) | |
Crystalline Powder | |
MFCD00013082 | |
14,2001 | |
Insoluble in water; Soluble in hot dilute HCl and H2SO4 | |
[Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O | |
544.29 | |
544.29 (Anhydrous) | |
Odorless | |
Cerium(III) oxalate hydrate |
15750-47-7 | |
C6Ce2O12 | |
UN3288 | |
cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate | |
ZMZNLKYXLARXFY-UHFFFAOYSA-H | |
cerium;oxalic acid;hydrate | |
91998046 | |
99% | |
Hygroscopic |
Safety and Handling
GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.