missing translation for 'onlineSavingsMsg'
Learn More
Cerium(IV) oxide, 99.5% (REO), Thermo Scientific Chemicals Product Code.: 11469850

Cerium(IV) oxide, 99.5% (REO), Thermo Scientific Chemicals

Product Code. 11469850
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11469850

Brand: Thermo Scientific Alfa Aesar 044662.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cerium(IV) oxide is used in ceramics, and to polish glass and stones. It is applied as solid electrolyte in fuel cells. It can also be employed as a co-catalyst in many number of reactions. It is also used as an antioxidant, and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles, and sunscreens in combination with other materials. It finds use as a catalyst support, and to sensitize photosensitive glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specificatie

Melting Point 2600°C
Color Yellow
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.5%
Solubility Information Insoluble in water.
Formula Weight 172.12
Density 7.132
Chemical Name or Material Cerium(IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN27K011 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number044662.22,044662.14,044662.36
Lot NumberZ03G017 Certificate TypeCertificate of Analysis Date19/09/2022 Catalog Number044662.22,044662.14,044662.36
Lot NumberU11H056 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044662.22,044662.14,044662.36
Lot NumberH01S033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044662.22,044662.14,044662.36
Lot NumberN25H058 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number044662.22,044662.14,044662.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium(IV) oxide, 99.5% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.