missing translation for 'onlineSavingsMsg'
Learn More

Cerium silicide, 99.9% (REO), Thermo Scientific Chemicals

95.78 € - 373.86 €

Chemical Identifiers

CAS 12014-85-6
Molecular Formula CeSi2
Molecular Weight (g/mol) 196.286
MDL Number MFCD00168082
InChI Key BLHYVSKDMXEWHD-UHFFFAOYSA-N
Synonym cerium silicide, cerium disilicide
PubChem CID 6336874
IUPAC Name bis($l^{2}-silanylidene)cerium
SMILES [Si]=[Ce]=[Si]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318737
View Documents
Thermo Scientific Alfa Aesar
035836.06
5 g
95.78 €
5g
Please to purchase this item. Need a web account? Register with us today!
11308737
View Documents
Thermo Scientific Alfa Aesar
035836.14
25 g
373.86 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12014-85-6
196.286
BLHYVSKDMXEWHD-UHFFFAOYSA-N
6336874
[Si]=[Ce]=[Si]
CeSi2
MFCD00168082
cerium silicide, cerium disilicide
bis($l^{2}-silanylidene)cerium

Specifications

1620°C
Gray
CeSi2
cerium silicide, cerium disilicide
BLHYVSKDMXEWHD-UHFFFAOYSA-N
bis($l^{2}-silanylidene)cerium
6336874
99.9%
5.31 g/mL
Cerium silicide
12014-85-6
Pieces
MFCD00168082
Insoluble in water.
[Si]=[Ce]=[Si]
196.286
196.29
Odorless
(REO)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-602-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.