Learn More
Abnova™ CES1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand:  Abnova™ H00001066-T01.100uL
Product Code. 16133121
  • 215.00 € / 100µL
Explore more special offers
Specifications
293T
CES1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 62.37kDa
Transient overexpression cell lysate was tested with anti-CES1 antibody (Mfr. No. H00001066-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
BC012418.1
pCMV-CES1 full-length
100 μL
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Abnova™ CES1 293T Cell Transient Overexpression Lysate (Denatured) > 100μL