Learn More
Cesium carbonate, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Cesium carbonate is widely utilized as a precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics and in the sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols and aerobic oxidation of alcohols into carbonyl compounds without polymeric by products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 610°C (decomposition) |
Color | White |
Physical Form | Powder/Granules |
Quantity | 10 g |
Assay Percent Range | 99.99% (Metal basis) |
Beilstein | 4546405 |
Solubility Information | Soluble in water,ethanol,dimethylformamide,dimethyl sulfoxide and methylpyrrolidone. |
IUPAC Name | dicaesium(1+) carbonate |
Formula Weight | 325.82 |
Odor | Odorless |
Show More |
Product Suggestions
Customers who viewed this item also viewed
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.