missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals
87.13 € - 378.89 €
Chemical Identifiers
CAS | 13587-19-4 |
---|---|
Molecular Formula | Cs2O4W |
MDL Number | MFCD00049438 |
Synonym | Cesium tungstate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11353598
|
Thermo Scientific Alfa Aesar
039206.06 |
5 g |
87.13 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11363598
|
Thermo Scientific Alfa Aesar
039206.14 |
25 g |
378.89 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cesium tungsten oxide is used for making transparent insulation coating that has the best infrared transparent barrier feature.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13587-19-4 | |
Cs2O4W | |
MFCD00049438 | |
Cesium tungstate |
Specifications
13587-19-4 | |
Powder | |
Cs2O4W | |
Cesium tungstate | |
513.66 | |
Hygroscopic |
White | |
99.9% (Metals basis) | |
MFCD00049438 | |
It is soluble in water. | |
Odorless | |
Cesium tungsten oxide |
Safety and Handling
missing translation for 'einecsNumber' : 237-019-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only