Learn More
Description
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Buffer; zwitterionic nondenaturing detergent for solubilizing membrane proteinsCHAPS is used in the laboratory to dissolve biological macromolecules such as proteins. It acts as a non-denaturing solvent used in purification of membrane proteins. It is also useful in conjunction with nonionic detergents like Triton X-100. Further, it is used as a non-denaturing zwitterionic detergent for membrane biochemistry.
Solubility
Slightly soluble in water.
Notes
Store in a cool. Incompatible with oxidizing agent, strong acid and bases.

Specifications
Specifications
Melting Point | 156°C to 158°C (decomposition) |
Quantity | 25 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 614.88 |
Percent Purity | ≥98% |
Chemical Name or Material | CHAPS |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253091 | Certificate of Analysis | 14/11/2024 | B21927.06,B21927.03,B21927.14 |
10251429 | Certificate of Analysis | 30/08/2024 | B21927.06,B21927.03,B21927.14 |
10249566 | Certificate of Analysis | 08/02/2024 | B21927.06,B21927.03,B21927.14 |
10218139 | Certificate of Analysis | 23/11/2021 | B21927.06,B21927.03,B21927.14 |
10233127 | Certificate of Analysis | 23/11/2021 | B21927.06,B21927.03,B21927.14 |
Safety and Handling

- CHAPS
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : CHAPS
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.