missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Charger 230V EU

Spare part charger 230 Volt EU for all new pipetus™ models and for opus™.

Brand:  Hirschmann™ 9907503

Product Code. 10156980

  • 125.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spare part charger 230 Volt EU for all new pipetus™ models and for opus™.
Specifications

Specifications

Charger
Battery 230V pipetus
230 V
230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ Charger 230V EU > For Use With: For use with battery 230V pipetus

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.