Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
Brand: Lovibond™ 147210
Description
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
- Compact and lightweight
- Provides the basis for a comprehensive and easy-to-use analysis system
- Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifications
Ammonia | |
Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions | |
0 to 1mg/L |
For Both Mobile and Static Water Analysis | |
1 Each | |
Checkit™ Comparator Single Parameter Test Kit |
Safety and Handling
- COMPARATOR LOVIBOND 147210 AMMONIA 0 TO 1MG/L
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H361d-Suspected of damaging the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : salicylic acid, sodium nitropruside dihydrate, lithium hydroxide monohydrate, sodium dichloroisocyan
Your input is important to us. Please complete this form to provide feedback related to the content on this product.