Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
Brand:  Lovibond™ 147130
 View more versions of this product
Product Code. 11725468
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
  • Compact and lightweight
  • Provides the basis for a comprehensive and easy-to-use analysis system
  • Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifications
Specifications
Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
1 Pack
Checkit™ Comparator Single Parameter Test Kit
30
4 to 10pH