Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
Brand: Lovibond™ 147026
This item is not returnable.
View return policy
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
pH value Chlorine |
|
6.5 to 8.4 pH 0 to 4 mg/L Cl2 |
|
Colorimetric | |
2 in 1 Test Kit |
Product Content Correction
Product Title
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits > pH and Chlorine 0 to 4 mg/L
Spot an opportunity for improvement?Share a Content Correction