Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
83.75 € - 106.00 €
Product Code | Brand | Description | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Range | Price | Quantity & Availability | |||||
10631613
|
Lovibond™
147045 |
pH value Chlorine |
6.5 to 8.4 pH 0.1 to 2 mg/L Cl2 |
N/A | ||||||
10739355
|
Lovibond™
147235 |
pH Value Copper |
6.5 to 8.4 pH 0 to 1 mg/L Cu |
N/A | ||||||
11735458
|
Lovibond™
147015 |
pH Value Chlorine |
6.5 to 8.4 pH 0 to 1 mg/L Cl2 |
N/A | ||||||
10193202
|
Lovibond™
147016 |
pH Value Chlorine |
6.5 to 8.4 pH 0 to 1 mg/L Cl2 |
96.80 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10739104
|
Lovibond™
147026 |
pH value Chlorine |
6.5 to 8.4 pH 0 to 4 mg/L Cl2 |
96.80 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10461045
|
Lovibond™
147285 |
pH Value Bromine |
6.5 to 8.4 pH 0 to 5 mg/L Br |
99.25 €
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Colorimetric | |
2 in 1 Test Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.