Learn More
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
Brand:  Lovibond™ 147015
 View more versions of this product
Product Code. 11735458
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitors 2-in-1 Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
  • Easy operation and exact reagent dosing
  • Tablet reagents with a minimm guaranteed shelf life of 5/10 years
  • Rotate continuous gradient color wheel to obtain a color match with the reacted sample
  • Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Specifications
pH Value
Chlorine
30
30 Tests
2 in 1 Test Kit
Checkit Discs, cells, stirring rod and Aqualytic tablet reagents (for 30 Tests) for the desired test
6.5 to 8.4 pH
0 to 1 mg/L Cl2
Colorimetric
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Checkit™ Comparators 2-in-1 Test Kits > pH and Chlorine 0 to 1 mg/L