Learn More
Lovibond™ Checkit™ Comparitor Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
Brand:  Lovibond™ 147370
 View more versions of this product
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10285022
  • 84.90 € / Each
Explore more special offers
Description
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
  • Easy operation and exact reagent dosing
  • Tablet reagents with a minimm guaranteed shelf life of 5/10 years
  • Rotate continuous gradient color wheel to obtain a color match with the reacted sample
  • Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Specifications
DEHA
0 to 0.5 mg/L
Colorimetric
Water test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Checkit™ Comparitor Test Kits > DEHA 0 to 0.5 mg/L