Learn More
Lovibond™ Checkit™ Comparitor Test Kits
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
77.10 € - 103.00 €
Product Code | Brand | Description | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Range | Price | Quantity & Availability | |||||
10285022
|
Lovibond™
147370 |
DEHA | 0 to 0.5 mg/L |
84.90 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10376712
|
Lovibond™
147330 |
Chlorine dioxide | 0.01 to 0.2 mg/L |
77.10 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
10708985
|
Lovibond™
147480 |
Phosphate, powder pack | 0 to 2.5 mg/L |
103.00 €
Each |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||||
Description
Perform water analysis quickly and easily. Lovibond Checkit™ Comparitor Test Kits are highly accurate and incorporate a user-friendly colorimetric scale. Each kit indicates a combination of pH and another measure.
- Easy operation and exact reagent dosing
- Tablet reagents with a minimm guaranteed shelf life of 5/10 years
- Rotate continuous gradient color wheel to obtain a color match with the reacted sample
- Printed continuous gradient color discs provide accurate measurements without "exrapolating" between discrete color blocks
Specifications
Colorimetric | |
Water test kit |
Safety and Handling
- testkit comparitor DEHA 0-0.5mg/l
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : nitric acid
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.