missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ BioPrepNylon™ Matrix Filters

Many biological preparations begin with extractions that require coarse filtration to remove cellular debris. Bulk filtration using cheesecloth absorbs much of the extraction fluid, while membranes clog quickly. BioPrepNylon™ produces rapid bulk filtration at extraordinary flow rates.

Brand:  Biodesign™ N301

Product Code. 12967487

  • 92.70 € / Pack of 3
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

BioPrepNylon produces rapid bulk filtration at extraordinary flow rates. BioPrepNylon is loosely placed in a sieve holder to form a funnel. The sample is placed in the funnel, allowed to drain, and the layers of the BioPrepNylon are gently pressed together to collect all of the extraction liquid.

  • Consists of very fine nylon filaments woven into a matrix with 50 micron pores.
  • A pure nylon product without detergents, dyes, or salts. Can be used directly with biologicals.
  • As BioPrepNylon does not wet, significantly greater quantities of the extraction liquid are recovered compared to any other method.
  • BioPrepNylon does not bind proteins, so no loss of your valuable sample will occur.
  • Compatible with the salts, buffers, and solvents used in biological research. Autoclavable.
  • Extraordinary cost effective compared to other bulk filtration methods. High tensile strength. Can be repeatedly washed and reused.
Specifications

Specifications

3 Pk.
Matrix Filter
Autoclavable
Square Sheet
50 μm
Nylon
30 x 30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Biodesign™ BioPrepNylon™ Matrix Filters > Pore Size: 50 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.